siRNA Details
| Gene Symbol: UFSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018359.3 | NP_060829.2 | 9, 10 | 1249 | ||
| NR_028085.1 | 9, 10 | 1224 | ||||
| GenBank mRNA | AK002062.1 | - | 9, 10 | 1178 | ||
| AK091635.1 | - | 9, 10 | 1217 | |||
| BC010493.1 | - | 8, 9 | 1145 | 3048997 | ||
| CR457278.1 | - | 8, 9 | 1132 | |||

UFSP2
Genomic Map