siRNA Details
| Gene Symbol: C19orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308277.1 | NP_001295206.1 | 3 | 483 | ||
| NM_018381.3 | NP_060851.2 | 3 | 483 | |||
| XM_011528121.1 | XP_011526423.1 | 3 | 483 | |||
| XM_011528122.1 | XP_011526424.1 | 3 | 483 | |||
| XM_017026934.1 | XP_016882423.1 | 2 | 153 | |||
| GenBank mRNA | AK001098.1 | - | 879 | |||
| AK002148.1 | - | 3 | 269 | |||
| AK096142.1 | - | 2 | 230 | |||
| AL137694.1 | - | 3 | 185 | |||
| BC035817.2 | - | 3 | 275 | 5554801 | ||

C19orf66
Genomic Map