siRNA Details
| Gene Symbol: LRRC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018509.3 | NP_060979.2 | 1 | 320 | ||
| GenBank mRNA | AK025328.1 | - | 1 | 320 | ||
| BC017168.1 | - | 1 | 278 | 3907072 | ||
| BC052279.1 | - | 1 | 238 | 4127952 | ||
| DA782636.1 | - | 1 | 320 | |||

LRRC59
Genomic Map