siRNA Details
Gene Symbol: GPALPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316951.1 | NP_001303880.1 | 8 | 1024 | ||
NM_001316952.1 | NP_001303881.1 | 8 | 1145 | |||
NM_018559.3 | NP_061029.2 | 8 | 1024 | |||
XM_005266444.3 | XP_005266501.1 | 8 | 1016 | |||
XM_011535144.2 | XP_011533446.1 | 8 | 1083 | |||
GenBank mRNA | AB051491.1 | - | 8 | 934 | ||
AF145122.1 | - | 8 | 959 | |||
AF245045.1 | - | 7 | 810 | |||
AK292493.1 | - | 8 | 997 | |||
BC032333.1 | - | 8 | 1071 | 5169953 | ||
BC039586.1 | - | 8 | 1002 | 5722236 |
