siRNA Details
| Gene Symbol: PRCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319214.1 | NP_001306143.1 | 3 | 409 | ||
| NM_005040.3 | NP_005031.1 | 4 | 771 | |||
| NM_199418.3 | NP_955450.2 | 5 | 834 | |||
| XM_005274093.1 | XP_005274150.1 | 4 | 501 | |||
| GenBank mRNA | AB451270.1 | - | 4 | 570 | ||
| AB451397.1 | - | 4 | 570 | |||
| AK091786.1 | - | 5 | 661 | |||
| AK302386.1 | - | 3 | 360 | |||
| AK312919.1 | - | 4 | 592 | |||
| BC001500.2 | - | 4 | 589 | 3510081 | ||
| L13977.1 | - | 4 | 599 | |||

PRCP
Genomic Map