siRNA Details
| Gene Symbol: PARL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037639.2 | NP_001032728.1 | 9 | 983 | ||
| NM_001324436.1 | NP_001311365.1 | 9 | 1031 | |||
| NM_001324437.1 | NP_001311366.1 | 8 | 881 | |||
| NM_001324438.1 | NP_001311367.1 | 8 | 933 | |||
| NM_018622.6 | NP_061092.3 | 10 | 1133 | |||
| NR_136893.1 | 10 | 1189 | ||||
| GenBank mRNA | AF197937.1 | - | 10 | 1071 | ||
| AK024765.1 | - | 10 | 1105 | |||
| BC003653.1 | - | 9 | 970 | 3537438 | ||
| BC014058.2 | - | 10 | 1091 | 3937238 | ||
| CA417847.1 | - | 1 | 290 | |||

PARL
Genomic Map