siRNA Details
| Gene Symbol: DDX43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018665.2 | NP_061135.2 | 15 | 2144 | ||
| XM_011535926.1 | XP_011534228.1 | 14 | 2012 | |||
| XM_011535927.2 | XP_011534229.1 | 14 | 1601 | |||
| XM_011535928.2 | XP_011534230.1 | 14 | 1790 | |||
| XM_017011003.1 | XP_016866492.1 | 14 | 1730 | |||
| GenBank mRNA | AJ278110.1 | - | 15 | 2007 | ||
| AK313686.1 | - | 15 | 1844 | |||
| AL136751.1 | - | 15 | 1832 | |||
| BC066938.1 | - | 15 | 1825 | 4829072 | ||
| BQ020439.1 | - | 3 | 560 | |||

DDX43
Genomic Map