siRNA Details
| Gene Symbol: SAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018666.2 | NP_061136.2 | 17 | 2189 | ||
| XM_017029621.1 | XP_016885110.1 | 17 | 2197 | |||
| XM_017029622.1 | XP_016885111.1 | 17 | 2108 | |||
| XM_017029623.1 | XP_016885112.1 | 17 | 2071 | |||
| XM_017029624.1 | XP_016885113.1 | 16 | 2011 | |||
| XM_017029626.1 | XP_016885115.1 | 11 | 1313 | |||
| XM_017029627.1 | XP_016885116.1 | 7 | 1104 | |||
| GenBank mRNA | AJ278111.1 | - | 17 | 2189 | ||
| AK310412.1 | - | 16 | 2145 | |||
| BC144260.1 | - | 10 | 942 | 9052780 | ||

SAGE1
Genomic Map