siRNA Details
| Gene Symbol: TRIM36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300752.1 | NP_001287681.1 | 2 | 664 | ||
| NM_001300759.1 | NP_001287688.1 | 3 | 683 | |||
| NM_018700.3 | NP_061170.2 | 3 | 899 | |||
| XM_017009621.1 | XP_016865110.1 | 5 | 911 | |||
| XM_017009622.1 | XP_016865111.1 | 5 | 1628 | |||
| XM_017009623.1 | XP_016865112.1 | 5 | 911 | |||
| GenBank mRNA | AB196558.1 | - | 2 | 155 | ||
| AJ272269.1 | - | 3 | 516 | |||
| AK296389.1 | - | 3 | 681 | |||
| AK316422.1 | - | 3 | 586 | |||
| BC130334.1 | - | 3 | 568 | 40146365 | ||

TRIM36
Genomic Map