siRNA Details
| Gene Symbol: CDC42BPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017525.2 | NP_059995.2 | 8 | 978 | ||
| XM_011545155.2 | XP_011543457.1 | 8 | 1093 | |||
| XM_011545156.2 | XP_011543458.1 | 8 | 1094 | |||
| XM_011545157.2 | XP_011543459.1 | 8 | 1094 | |||
| XM_011545158.2 | XP_011543460.1 | 8 | 1094 | |||
| XM_011545159.2 | XP_011543461.1 | 8 | 1094 | |||
| XM_011545160.2 | XP_011543462.1 | 9 | 1045 | |||
| XM_011545161.2 | XP_011543463.1 | 8 | 1094 | |||
| XM_017017996.1 | XP_016873485.1 | 8 | 1093 | |||
| XM_017017997.1 | XP_016873486.1 | 8 | 1094 | |||
| XM_017017998.1 | XP_016873487.1 | 8 | 1094 | |||
| GenBank mRNA | AY648038.1 | - | 8 | 978 | ||
| Y12337.1 | - | 5 | 642 | |||

CDC42BPG
Genomic Map