siRNA Details
| Gene Symbol: RNPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017619.3 | NP_060089.1 | 8, 9 | 1012 | ||
| GenBank mRNA | AK000015.1 | - | 2, 3 | 187 | ||
| AK057799.1 | - | 8, 9 | 991 | |||
| AY099329.1 | - | 8, 9 | 997 | |||
| BC010697.1 | - | 8, 9 | 988 | 3873751 | ||

RNPC3
Genomic Map