siRNA Details
| Gene Symbol: TTC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193509.1 | NP_001180438.1 | 11, 12 | 1359 | ||
| NM_017735.4 | NP_060205.3 | 11, 12 | 1606 | |||
| XM_011532958.1 | XP_011531260.1 | 11, 12 | 1593 | |||
| GenBank mRNA | AK000279.1 | - | 8, 9 | 814 | ||
| AK000665.1 | - | 6, 7 | 598 | |||
| AK027570.1 | - | 11, 12 | 1515 | |||
| AK225295.1 | - | 8, 9 | 814 | |||
| AK299196.1 | - | 11, 12 | 1348 | |||
| BC001248.1 | - | 10, 11 | 1139 | |||
| BC063791.1 | - | 11, 12 | 1564 | 6500951 | ||

TTC27
Genomic Map