siRNA Details
| Gene Symbol: G2E3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308097.1 | NP_001295026.1 | 5 | 551 | ||
| NM_017769.4 | NP_060239.2 | 6 | 592 | |||
| XM_011536925.2 | XP_011535227.1 | 6 | 558 | |||
| XM_011536926.1 | XP_011535228.1 | 6 | 592 | |||
| GenBank mRNA | AB037754.1 | - | 6 | 544 | ||
| AK000340.1 | - | 6 | 557 | |||
| AK093536.1 | - | 6 | 1579 | |||
| AK225346.1 | - | 6 | 557 | |||
| AK295571.1 | - | 5 | 516 | |||
| AK296872.1 | - | 5 | 516 | |||
| BC000973.2 | - | 6 | 538 | 3450488 | ||

G2E3
Genomic Map