siRNA Details
| Gene Symbol: KRBOX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129898.1 | NP_001123370.1 | 6 | 664 | ||
| NM_001129899.1 | NP_001123371.1 | 7 | 708 | |||
| NM_001129900.1 | NP_001123372.1 | 7 | 693 | |||
| NM_017776.2 | NP_060246.2 | 6 | 649 | |||
| GenBank mRNA | AK000351.1 | - | 6 | 514 | ||
| AK096413.1 | - | 6 | 556 | |||
| AK097159.1 | - | 1 | 1538 | |||
| AK289703.1 | - | 6 | 546 | |||
| BC012569.1 | - | 6 | 516 | 4283420 | ||

KRBOX4
Genomic Map