siRNA Details
| Gene Symbol: FLVCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195283.1 | NP_001182212.1 | 8 | 873 | ||
| NM_017791.2 | NP_060261.2 | 8 | 1755 | |||
| GenBank mRNA | AF456126.1 | - | 8 | 1379 | ||
| AK000378.1 | - | 8 | 1703 | |||
| AK027804.1 | - | 7 | 643 | |||
| AK297002.1 | - | 8 | 873 | |||
| AY260572.1 | - | 8 | 1735 | |||
| AY260577.1 | - | 8 | 1735 | |||
| BC019087.2 | - | 8 | 1718 | 4866427 | ||

FLVCR2
Genomic Map