siRNA Details
| Gene Symbol: BCAS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010974.2 | NP_001010974.1 | 4 | 505 | ||
| NM_017843.4 | NP_060313.3 | 6 | 737 | |||
| NM_198799.3 | NP_942094.2 | 5 | 640 | |||
| XM_011528886.2 | XP_011527188.1 | 6 | 742 | |||
| XM_011528887.2 | XP_011527189.1 | 5 | 563 | |||
| GenBank mRNA | AF361220.1 | - | 6 | 737 | ||
| AJ511266.1 | - | 4 | 329 | |||
| AK000502.1 | - | 6 | 600 | |||
| BC038381.1 | - | 5 | 490 | 4472674 | ||
| BC047337.1 | - | 6 | 566 | |||

BCAS4
Genomic Map