siRNA Details
| Gene Symbol: TMEM127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193304.2 | NP_001180233.1 | 4 | 738 | ||
| NM_017849.3 | NP_060319.1 | 4 | 761 | |||
| XM_017004450.1 | XP_016859939.1 | 3 | 342 | |||
| XM_017004451.1 | XP_016859940.1 | 3 | 387 | |||
| XM_017004452.1 | XP_016859941.1 | 3 | 342 | |||
| GenBank mRNA | AK000514.1 | - | 4 | 742 | ||
| AK293276.1 | - | 6 | 642 | |||
| BC028575.1 | - | 4 | 719 | 4837629 | ||
| BC039892.1 | - | 4 | 714 | 5725347 | ||

TMEM127
Genomic Map