siRNA Details
| Gene Symbol: NLRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174081.1 | NP_001167552.1 | 13 | 3237 | ||
| NM_001174082.1 | NP_001167553.1 | 12 | 3202 | |||
| NM_001174083.1 | NP_001167554.1 | 14 | 3199 | |||
| NM_017852.3 | NP_060322.1 | 13 | 3268 | |||
| GenBank mRNA | AB209300.1 | - | 12 | 3118 | ||
| AF269288.2 | - | 3 | 1469 | |||
| AF298547.1 | - | 12 | 3116 | |||
| AF310106.1 | - | 13 | 3213 | |||
| AF464764.1 | - | 13 | 3168 | |||
| AK000517.1 | - | 13 | 3213 | |||
| AK000784.1 | - | 8 | 1388 | |||
| AK025952.1 | - | 13 | 3177 | |||
| AK223253.1 | - | 13 | 3177 | |||
| AK223269.1 | - | 13 | 3177 | |||
| AK292681.1 | - | 13 | 3196 | |||
| AK302989.1 | - | 14 | 3168 | |||
| BC001039.2 | - | 12 | 3148 | 3504594 | ||
| BC003592.1 | - | 13 | 3192 | 2821428 | ||
| BC039269.1 | - | 13 | 3438 | 5298283 | ||

NLRP2
Genomic Map