siRNA Details
| Gene Symbol: MREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018000.2 | NP_060470.2 | 2 | 509 | ||
| XM_011511468.2 | XP_011509770.1 | 2 | 359 | |||
| XM_017004475.1 | XP_016859964.1 | 2 | 359 | |||
| GenBank mRNA | AK000978.1 | - | 2 | 479 | ||
| BC032747.1 | - | 2 | 518 | 5573149 | ||
| BC082990.1 | - | 2 | 499 | 6159624 | ||
| BQ928130.1 | - | 2 | 509 | |||
| CX781801.1 | - | 2 | 504 | |||
| Gene Symbol: PECR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK296208.1 | - | 8 | 1081 | ||

MREG
Genomic Map