siRNA Details
| Gene Symbol: RBM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018047.2 | NP_060517.1 | 6 | 554 | ||
| GenBank mRNA | AK001152.1 | - | 6 | 509 | ||
| AK297019.1 | - | 5 | 323 | |||
| AL136933.1 | - | 6 | 483 | |||
| BC003402.1 | - | 6 | 487 | 3449258 | ||
| DA534416.1 | - | 6 | 555 | |||

RBM22
Genomic Map