siRNA Details
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318716.1 | NP_001305645.1 | 2 | 394 | ||
NM_001318717.1 | NP_001305646.1 | 2 | 315 | |||
NM_001318718.1 | NP_001305647.1 | 2 | 295 | |||
NM_001318719.1 | NP_001305648.1 | 2 | 273 | |||
NM_001318720.1 | NP_001305649.1 | 2 | 267 | |||
NM_001318721.1 | NP_001305650.1 | 2 | 242 | |||
NM_001318722.1 | NP_001305651.1 | 2 | 263 | |||
NM_001318723.1 | NP_001305652.1 | 2 | 347 | |||
NM_001318724.1 | NP_001305653.1 | 2 | 347 | |||
NM_001318725.1 | NP_001305654.1 | 2 | 250 | |||
NM_016506.6 | NP_057590.3 | 2 | 351 | |||
NM_018095.5 | NP_060565.4 | 2 | 246 | |||
XM_017018009.1 | XP_016873498.1 | 2 | 230 | |||
GenBank mRNA | AF151086.1 | - | 2 | 208 | ||
AK001312.1 | - | 2 | 200 | |||
AK001749.1 | - | 2 | 287 | |||
AK091607.1 | - | 1 | 924 | |||
AK222550.1 | - | 2 | 197 | |||
AK222985.1 | - | 2 | 207 | |||
AK225062.1 | - | 2 | 197 | |||
AL713735.1 | - | 1 | 176 | |||
BC002736.2 | - | 2 | 288 | 3628290 | ||
BI601113.1 | - | 2 | 254 | |||
BI908078.1 | - | 2 | 253 | |||
CR457270.1 | - | 1 | 134 | |||
DA016320.1 | - | 2 | 246 | |||
DA170281.1 | - | 2 | 302 | |||
DA559313.1 | - | 2 | 305 | |||
DA883418.1 | - | 2 | 284 | |||
DB457928.1 | - | 2 | 218 | |||
HY062454.1 | - | 2 | 229 | |||
HY120885.1 | - | 2 | 228 | |||
HY176105.1 | - | 2 | 270 | |||
HY182004.1 | - | 2 | 378 |
