siRNA Details
| Gene Symbol: LMBR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300750.1 | NP_001287679.1 | 9 | 1222 | ||
| NM_001300751.1 | NP_001287680.1 | 8 | 1020 | |||
| NM_018113.3 | NP_060583.2 | 8 | 1020 | |||
| XM_011538563.2 | XP_011536865.1 | 8 | 1174 | |||
| XM_011538564.2 | XP_011536866.1 | 8 | 1547 | |||
| XM_011538565.2 | XP_011536867.1 | 7 | 1796 | |||
| XM_017019626.1 | XP_016875115.1 | 8 | 1422 | |||
| XM_017019627.1 | XP_016875116.1 | 9 | 1211 | |||
| XM_017019628.1 | XP_016875117.1 | 8 | 1009 | |||
| XM_017019629.1 | XP_016875118.1 | 9 | 2066 | |||
| GenBank mRNA | AB033000.1 | - | 8 | 787 | ||
| AF260728.1 | - | 8 | 979 | |||
| AK001356.1 | - | 9 | 1003 | |||
| AK001651.1 | - | 8 | 970 | |||
| AK093570.1 | - | 3 | 806 | |||
| AK298859.1 | - | 2 | 74 | |||
| AK310064.1 | - | 9 | 1156 | |||
| AL137599.1 | - | 5 | 413 | |||
| AY358572.1 | - | 8 | 984 | |||
| BC008337.1 | - | 8 | 950 | 3506347 | ||
| BC015015.1 | - | 8 | 749 | 3910520 | ||
| BC031550.1 | - | 8 | 781 | 5194023 | ||
| KU178711.1 | - | 8 | 675 | |||
| KU178712.1 | - | 7 | 658 | |||
| KU178713.1 | - | 7 | 641 | |||
| KU178714.1 | - | 8 | 675 | |||
| KU178715.1 | - | 8 | 675 | |||

LMBR1L
Genomic Map