siRNA Details
| Gene Symbol: TMEM184C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018241.2 | NP_060711.2 | 6, 7 | 1233 | ||
| GenBank mRNA | AK000169.1 | - | 3, 4 | 386 | ||
| AK001708.1 | - | 6, 7 | 1176 | |||
| BC046128.1 | - | 6, 7 | 1235 | 5750613 | ||

TMEM184C
Genomic Map