siRNA Details
| Gene Symbol: TMEM30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018247.3 | NP_060717.1 | 2, 3 | 626 | ||
| GenBank mRNA | AK001718.1 | - | 2, 3 | 495 | ||
| AK293225.1 | - | 3, 4 | 451 | |||
| AK295567.1 | - | 3, 4 | 447 | |||
| AL832490.1 | - | 2, 3 | 549 | |||
| AL832815.1 | - | 2, 3 | 488 | |||

TMEM30A
Genomic Map