siRNA Details
| Gene Symbol: H2AFJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177925.3 | NP_808760.1 | 1 | 162 | ||
| NR_027716.2 | 1 | 162 | ||||
| GenBank mRNA | AK001765.1 | - | 1 | 93 | ||
| BC003602.1 | - | 1 | 83 | 3354610 | ||
| BM689354.1 | - | 1 | 166 | |||
| BU594879.1 | - | 1 | 140 | |||

H2AFJ
Genomic Map