siRNA Details
| Gene Symbol: MBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018328.4 | NP_060798.2 | 9 | 3335 | ||
| XM_011511470.2 | XP_011509772.1 | 9 | 3567 | |||
| XM_011511472.2 | XP_011509774.1 | 7 | 3147 | |||
| XM_017004482.1 | XP_016859971.1 | 5 | 3017 | |||
| XM_017004483.1 | XP_016859972.1 | 5 | 3017 | |||
| XM_017004484.1 | XP_016859973.1 | 5 | 3017 | |||
| XM_017004485.1 | XP_016859974.1 | 5 | 3017 | |||
| GenBank mRNA | AB040894.1 | - | 4 | 2628 | ||
| AK304424.1 | - | 4 | 2650 | |||
| BC014534.1 | - | 1 | 784 | |||
| BC150264.1 | - | 4 | 2628 | 9007212 | ||
| EF542797.1 | - | 9 | 3326 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001290656, NM_029924, AK173176, BQ551845, BC138375, and others.

MBD5
Genomic Map