siRNA Details
| Gene Symbol: EMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018447.3 | NP_060917.1 | 3, 4 | 683 | ||
| XM_005265321.3 | XP_005265378.1 | 3, 4 | 551 | |||
| GenBank mRNA | AF157321.1 | - | 2, 3 | 395 | ||
| AK172843.1 | - | 2, 3 | 391 | |||
| AK222953.1 | - | 2, 3 | 393 | |||
| AK312008.1 | - | 2, 3 | 390 | |||
| BC022807.1 | - | 2, 3 | 378 | 4897577 | ||
| DA359966.1 | - | 3, 4 | 551 | |||

EMC3
Genomic Map