siRNA Details
| Gene Symbol: EAPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318916.1 | NP_001305845.1 | 4 | 518 | ||
| NM_018453.3 | NP_060923.2 | 5 | 636 | |||
| XM_011536958.1 | XP_011535260.1 | 5 | 914 | |||
| XM_017021460.1 | XP_016876949.1 | 5 | 636 | |||
| GenBank mRNA | AK000585.1 | - | 5 | 600 | ||
| AK225467.1 | - | 5 | 591 | |||
| AK225477.1 | - | 5 | 600 | |||
| AK225991.1 | - | 5 | 600 | |||
| AY869694.1 | - | 5 | 559 | |||
| BC001245.1 | - | 5 | 609 | 3460193 | ||
| BU662131.1 | - | 4 | 466 | |||

EAPP
Genomic Map