siRNA Details
| Gene Symbol: WWC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015691.3 | NP_056506.2 | 3 | 272 | ||
| GenBank mRNA | AI306162.1 | - | 4 | 336 | ||
| AK292961.1 | - | 2 | 154 | |||
| BU500300.1 | - | 3 | 480 | |||
| BU500742.1 | - | 3 | 456 | |||
| KC987947.1 | - | 3 | 449 | |||

WWC3
Genomic Map