siRNA Details
| Gene Symbol: CMAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018686.5 | NP_061156.1 | 8 | 1332 | ||
| NR_135117.1 | 7 | 1160 | ||||
| GenBank mRNA | AF271388.1 | - | 8 | 1220 | ||
| AF397212.1 | - | 8 | 1202 | |||
| AK022927.1 | - | 8 | 1242 | |||
| AK124990.1 | - | 1 | 1674 | |||
| AK222981.1 | - | 8 | 1231 | |||
| AL832975.1 | - | 8 | 1280 | |||
| BC016609.1 | - | 7 | 1109 | 3448277 | ||
| BM671960.1 | - | 1 | 474 | |||

CMAS
Genomic Map