siRNA Details
| Gene Symbol: GNG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018841.5 | NP_061329.3 | 4 | 312 | ||
| XM_017001809.1 | XP_016857298.1 | 4 | 479 | |||
| XM_017001810.1 | XP_016857299.1 | 5 | 720 | |||
| XM_017001811.1 | XP_016857300.1 | 5 | 649 | |||
| GenBank mRNA | AF119663.1 | - | 4 | 296 | ||
| AF123766.1 | - | 4 | 303 | |||
| AF365871.1 | - | 4 | 239 | |||
| AF493879.1 | - | 2 | 107 | |||
| AK223000.1 | - | 4 | 254 | |||
| AK293101.1 | - | 5 | 253 | |||
| AL832431.1 | - | 4 | 265 | |||
| BC005940.1 | - | 4 | 293 | 4049838 | ||
| BP236225.1 | - | 4 | 299 | |||
| BP236699.1 | - | 4 | 312 | |||

GNG12
Genomic Map