siRNA Details
Gene Symbol: SMG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005259057.2 | XP_005259114.1 | 12 | 1652 | ||
XM_011527113.1 | XP_011525415.1 | 12 | 1407 | |||
XM_011527114.1 | XP_011525416.1 | 12 | 1345 | |||
XM_011527115.1 | XP_011525417.1 | 12 | 1413 | |||
XM_011527116.1 | XP_011525418.1 | 12 | 1539 | |||
XM_017026988.1 | XP_016882477.1 | 8 | 763 | |||
GenBank mRNA | AK022948.1 | - | 12 | 1624 | ||
AL136606.1 | - | 12 | 1578 | |||
BC008869.2 | - | 12 | 1564 | 4131601 |
