siRNA Details
| Gene Symbol: TDRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198795.1 | NP_942090.1 | 19 | 2805 | ||
| XM_005269978.2 | XP_005270035.1 | 19 | 2786 | |||
| XM_005269979.2 | XP_005270036.1 | 19 | 2819 | |||
| XM_011539959.2 | XP_011538261.1 | 19 | 3158 | |||
| XM_011539960.2 | XP_011538262.1 | 19 | 2748 | |||
| XM_011539961.2 | XP_011538263.1 | 19 | 2730 | |||
| XM_011539962.1 | XP_011538264.1 | 19 | 3112 | |||
| XM_011539963.1 | XP_011538265.1 | 19 | 2819 | |||
| XM_011539964.1 | XP_011538266.1 | 18 | 2648 | |||
| XM_011539966.1 | XP_011538268.1 | 18 | 2648 | |||
| XM_017016414.1 | XP_016871903.1 | 18 | 2675 | |||
| XM_017016415.1 | XP_016871904.1 | 17 | 2504 | |||
| GenBank mRNA | AF285606.1 | - | 12 | 1774 | ||
| AK024735.1 | - | 9 | 1325 | |||
| AK122916.1 | - | 19 | 2815 | |||
| AK304331.1 | - | 10 | 1602 | |||
| AL833340.1 | - | 19 | 2807 | |||
| BC035010.2 | - | 19 | 3111 | |||
| BC063133.1 | - | 17 | 2371 | 6710928 | ||
| BX648664.1 | - | 19 | 2751 | |||

TDRD1
Genomic Map