siRNA Details
| Gene Symbol: ZNF253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021047.2 | NP_066385.2 | 4 | 1321 | ||
| GenBank mRNA | AK074757.1 | - | 5 | 1229 | ||
| AK074872.1 | - | 1 | 971 | |||
| BC014148.2 | - | 4 | 1321 | |||
| BC065572.1 | - | 2 | 1096 | 6171141 | ||
| BC125063.1 | - | 4 | 1300 | 40117279 | ||
| BC125064.1 | - | 2 | 1077 | 40117289 | ||

ZNF253
Genomic Map