siRNA Details
| Gene Symbol: YLPM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019589.2 | NP_062535.2 | 13 | 5731 | ||
| XM_005267860.3 | XP_005267917.1 | 13 | 5775 | |||
| XM_011536966.2 | XP_011535268.1 | 14 | 4890 | |||
| XM_011536967.1 | XP_011535269.1 | 12 | 3657 | |||
| GenBank mRNA | AK090435.1 | - | 11 | 4487 | ||
| AK297582.1 | - | 9 | 1302 | |||
| AL832365.1 | - | 10 | 4266 | |||
| BC007792.1 | - | 4 | 328 | |||
| L40403.1 | - | 7 | 834 | |||

YLPM1
Genomic Map