siRNA Details
| Gene Symbol: DNAJC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021800.2 | NP_068572.1 | 3 | 404 | ||
| NM_201262.1 | NP_957714.1 | 3 | 404 | |||
| XM_017016431.1 | XP_016871920.1 | 2 | 176 | |||
| XM_017016432.1 | XP_016871921.1 | 2 | 147 | |||
| GenBank mRNA | AF176012.1 | - | 3 | 389 | ||
| AF176013.1 | - | 3 | 321 | |||
| AK289537.1 | - | 3 | 354 | |||
| BC017018.1 | - | 3 | 340 | 3884093 | ||
| CR457184.1 | - | 3 | 236 | |||

DNAJC12
Genomic Map