siRNA Details
| Gene Symbol: RETN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193374.1 | NP_001180303.1 | 4 | 303 | ||
| NM_020415.3 | NP_065148.1 | 4 | 312 | |||
| GenBank mRNA | AB111910.1 | - | 2 | 188 | ||
| AF205952.1 | - | 3 | 290 | |||
| AF290874.1 | - | 4 | 312 | |||
| AF323081.1 | - | 4 | 312 | |||
| AY207314.1 | - | 4 | 303 | |||
| AY359066.1 | - | 3 | 290 | |||
| BC069302.1 | - | 4 | 312 | 7262685 | ||
| BC101554.1 | - | 4 | 295 | 8069060 | ||
| BC101560.1 | - | 4 | 295 | 8069066 | ||

RETN
Genomic Map