siRNA Details
| Gene Symbol: PSMA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002787.4 | NP_002778.1 | 8 | 669 | ||
| GenBank mRNA | AK222976.1 | - | 8 | 637 | ||
| AK290654.1 | - | 8 | 644 | |||
| AU120930.1 | - | 8 | 644 | |||
| BC002900.2 | - | 7 | 595 | |||
| BC047697.1 | - | 8 | 623 | 5786518 | ||
| CR450317.1 | - | 8 | 621 | |||
| D00760.1 | - | 8 | 621 | |||

PSMA2
Genomic Map