siRNA Details
| Gene Symbol: PSMA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002787.4 | NP_002778.1 | 1, 2 | 87 | ||
| GenBank mRNA | AK222976.1 | - | 1, 2 | 55 | ||
| AK290654.1 | - | 1, 2 | 62 | |||
| AU120930.1 | - | 1, 2 | 62 | |||
| BC002900.2 | - | 13 | ||||
| BC047697.1 | - | 1, 2 | 41 | 5786518 | ||
| CB990982.1 | - | 1, 2 | 130 | |||
| CR450317.1 | - | 1, 2 | 39 | |||
| D00760.1 | - | 1, 2 | 39 | |||

PSMA2
Genomic Map