siRNA Details
| Gene Symbol: UBQLN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304342.1 | NP_001291271.1 | 11 | 1767 | ||
| NM_020131.4 | NP_064516.2 | 11 | 1827 | |||
| GenBank mRNA | AB209566.1 | - | 11 | 1700 | ||
| AF113544.1 | - | 8 | 1233 | |||
| AF188240.1 | - | 11 | 1981 | |||
| AK098368.1 | - | 6 | 672 | |||
| AK302895.1 | - | 11 | 1656 | |||
| AK314413.1 | - | 11 | 1715 | |||
| BC006410.1 | - | 2 | 102 | |||
| BC018403.2 | - | 11 | 1715 | 4214386 | ||
| BC063841.1 | - | 11 | 1727 | 6183942 | ||

UBQLN4
Genomic Map