siRNA Details
| Gene Symbol: OTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020157.3 | NP_064542.1 | 2 | 188 | ||
| XM_017027959.1 | XP_016883448.1 | 2 | 164 | |||
| GenBank mRNA | AF233261.1 | - | 2 | 183 | ||
| AF243505.1 | - | 2 | 146 | |||
| AJ242552.1 | - | 2 | 139 | |||
| AY359082.1 | - | 2 | 176 | |||
| BC101688.1 | - | 2 | 188 | 8069194 | ||
| BC101690.1 | - | 2 | 188 | 8069196 | ||

OTOR
Genomic Map