siRNA Details
| Gene Symbol: STOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020225.2 | NP_064610.1 | 3 | 2688 | ||
| XM_011532129.1 | XP_011530431.1 | 3 | 2812 | |||
| XM_011532130.1 | XP_011530432.1 | 3 | 2812 | |||
| XM_011532131.1 | XP_011530433.1 | 6 | 1943 | |||
| XM_017008466.1 | XP_016863955.1 | 3 | 2812 | |||
| XM_017008467.1 | XP_016863956.1 | 4 | 1849 | |||
| GenBank mRNA | AB037813.1 | - | 3 | 2688 | ||
| AK289840.1 | - | 3 | 1367 | |||
| BC146754.1 | - | 3 | 2688 | 9007190 | ||

STOX2
Genomic Map