siRNA Details
| Gene Symbol: PSMB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002801.3 | NP_002792.1 | 4 | 434 | ||
| GenBank mRNA | AK225658.1 | - | 4 | 407 | ||
| AK298012.1 | - | 2 | 742 | |||
| AK312208.1 | - | 4 | 394 | |||
| BC052369.1 | - | 4 | 372 | 4054165 | ||
| BM981173.1 | - | 5 | 582 | |||
| BP289065.1 | - | 4 | 434 | |||
| BT019723.1 | - | 4 | 306 | |||
| Y13640.1 | - | 4 | 394 | |||

PSMB10
Genomic Map