siRNA Details
| Gene Symbol: PITHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020362.4 | NP_065095.2 | 1, 2 | 329 | ||
| GenBank mRNA | AF218024.1 | - | 1, 2 | 310 | ||
| AF221595.1 | - | 1, 2 | 321 | |||
| AF271784.1 | - | 1, 2 | 325 | |||
| AK313005.1 | - | 1, 2 | 297 | |||
| BC017208.2 | - | 1, 2 | 290 | 3943360 | ||
| DB104988.1 | - | 1, 2 | 329 | |||

PITHD1
Genomic Map