siRNA Details
| Gene Symbol: C12orf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304811.1 | NP_001291740.1 | 2 | 213 | ||
| NM_020374.3 | NP_065107.1 | 2 | 221 | |||
| XM_005253716.1 | XP_005253773.1 | 2 | 221 | |||
| XM_006718992.3 | XP_006719055.1 | 2 | 221 | |||
| GenBank mRNA | AJ272205.1 | - | 2 | 164 | ||
| AK074479.1 | - | 2 | 221 | |||
| AL834377.1 | - | 2 | 184 | |||
| BC016908.1 | - | 2 | 164 | 4402263 | ||
| BX641128.1 | - | 2 | 214 | |||
| DA195770.1 | - | 2 | 186 | |||

C12orf4
Genomic Map