siRNA Details
| Gene Symbol: ZNF695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020394.4 | NP_065127.4 | 4 | 597 | ||
| GenBank mRNA | CN394881.1 | - | 4 | 529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF695| Gene Symbol: ZNF695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020394.4 | NP_065127.4 | 4 | 597 | ||
| GenBank mRNA | CN394881.1 | - | 4 | 529 | ||