siRNA Details
| Gene Symbol: RNPEPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018226.4 | NP_060696.4 | 7 | 1307 | ||
| XM_005247036.4 | XP_005247093.1 | 7 | 1368 | |||
| GenBank mRNA | AF300795.1 | - | 7 | 1013 | ||
| AK001668.1 | - | 3 | 271 | |||
| AK130805.1 | - | 2 | 958 | |||
| BC017301.2 | - | 4 | 459 | |||
| BC067258.1 | - | 7 | 882 | 6183046 | ||
| BC082975.1 | - | 7 | 1207 | 6379537 | ||
| BC099906.1 | - | 5 | 1420 | 5311883 | ||

RNPEPL1
Genomic Map