siRNA Details
| Gene Symbol: TMEM63C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020431.3 | NP_065164.2 | 15, 16 | 1561 | ||
| GenBank mRNA | AK124749.1 | - | 1, 2 | 69 | ||
| AK125159.1 | - | 15, 16 | 1493 | |||
| BC136613.1 | - | 15, 16 | 1443 | 9020603 | ||
| BC136614.1 | - | 15, 16 | 1440 | 9020604 | ||

TMEM63C
Genomic Map