siRNA Details
| Gene Symbol: LRTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304389.1 | NP_001291318.1 | 2 | 526 | ||
| NM_020678.3 | NP_065729.1 | 2 | 535 | |||
| GenBank mRNA | AF225421.1 | - | 2 | 506 | ||
| BC040732.1 | - | 2 | 526 | 5223743 | ||
| BC074741.2 | - | 2 | 396 | 30915199 | ||
| BM716618.1 | - | 2 | 535 | |||

LRTM1
Genomic Map