siRNA Details
| Gene Symbol: UGT2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105677.2 | NP_001099147.2 | 1 | 529 | ||
| NM_001301233.1 | NP_001288162.1 | 1 | 529 | |||
| GenBank mRNA | FJ664272.1 | - | 1 | 529 | ||
| FJ664273.1 | - | 1 | 529 | |||
| Gene Symbol: UGT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252274.2 | NP_001239203.2 | 3 | 1246 | ||
| GenBank mRNA | AK304249.1 | - | 3 | 1246 | ||

UGT2A2
Genomic Map